Methyldiphenylphosphine ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: Diphenylmethylphosphine

H27159.03 H27159.09 H27159.18
ALFAH27159.03EA 54.6 SGD
ALFAH27159.03 ALFAH27159.09 ALFAH27159.18
Methyldiphenylphosphine ≥99%
Methyldiphenylphosphine

Purity: 99% CAS No.: 1486-28-8 Molecular Formula: (C6H5)2PCH3 Formula Weight: 200.21 Flash Point: 110°(230°F) Boiling Point: 160°/15mm MDL No.: MFCD00008508

Formula: C₁₃H₁₃P
MW: 200,22 g/mol
Boiling Pt: 160 °C (15 mmHg)
Melting Pt: 28 °C
Density: 1,064
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008508
CAS Number: 1486-28-8
EINECS: 216-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR