1,2,3,4,5-Pentaphenylcyclopentadiene ≥99%

Supplier: Thermo Scientific

Synonyms: 1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene, Cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, Pentaphenylcyclopentadiene

H27223.MD H27223.06 H27223.03
ALFAH27223.MDEA 0 SGD
ALFAH27223.MD ALFAH27223.06 ALFAH27223.03
1,2,3,4,5-Pentaphenylcyclopentadiene ≥99%
1,2,3,4,5-Pentaphenylcyclopentadiene

Purity: 99% CAS No.: 2519-10-0 Molecular Formula: C35H26 Formula Weight: 446.59 Melting Point: 254-257° MDL No.: MFCD09909572

Formula: C₃₅H₂₆
MW: 446,59 g/mol
Melting Pt: 254…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00012263
CAS Number: 2519-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR