4,6-Dibromopyrimidine 95%
Supplier: Thermo Scientific
|
Danger
|
H27254.06
H27254.03
ALFAH27254.06EA
0
SGD
ALFAH27254.06
ALFAH27254.03
4,6-Dibromopyrimidine 95%
4,6-Dibromopyrimidine
Purity: 95%
CAS No.: 36847-10-6
Molecular Formula: C4H2Br2N2
Formula Weight: 237.88
Melting Point: 49-52°
UN No.: 3261
MDL No.: MFCD00233946
Formula:
C₄H₂Br₂N₂ Melting Pt: 49…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233946 CAS Number: 36847-10-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...