1-Hexyl-3-methylimidazolium tetrafluoroborate ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 1-n-Hexyl-3-methylimidazolium tetrafluoroborate, [HMIM] [BF4]

H27548.06 H27548.18
ALFAH27548.06EA 0 SGD
ALFAH27548.06 ALFAH27548.18
1-Hexyl-3-methylimidazolium tetrafluoroborate ≥99%
1-Hexyl-3-methylimidazolium tetrafluoroborate

Purity: 99% CAS No.: 244193-50-8 Molecular Formula: C10H19BF4N2 Formula Weight: 254.08 Refractive Index: 1.4300 Melting Point: -81° MDL No.: MFCD03095459

Formula: C₁₀H₁₉BF₄N₂
MW: 254,08 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: –81 °C
Density: 1,149
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD03095459
CAS Number: 244193-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR