6-Fluoropyridine-2-carbonitrile 95%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Fluoropicolinonitrile, 2-Cyano-6-fluoropyridine

H27600.MD H27600.03 H27600.06
ALFAH27600.MDEA 0 SGD
ALFAH27600.MD ALFAH27600.03 ALFAH27600.06
6-Fluoropyridine-2-carbonitrile 95%
6-Fluoropyridine-2-carbonitrile

Purity: 95% CAS No.: 3939-15-9 Molecular Formula: C6H3FN2 Formula Weight: 122.10 Melting Point: 32-35° MDL No.: MFCD09263323

Formula: C₆H₃FN₂
MW: 122,1 g/mol
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD09263323
CAS Number: 3939-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR