5-Bromoindol-3-ylacetic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Bromoindole-3-acetic acid, 2-(5-Bromo-1H-indol-3-yl)acetic acid

H29217.MD H29217.03
ALFAH29217.MDEA 0 SGD
ALFAH29217.MD ALFAH29217.03
5-Bromoindol-3-ylacetic acid ≥97%
5-Bromoindol-3-ylacetic acid

Purity: 97% CAS No.: 40432-84-6 Molecular Formula: C10H8BrNO2 Formula Weight: 254.09 Melting Point: 144-145° dec. MDL No.: MFCD00005637

Formula: C₁₀H₈BrNO₂
MW: 254,08 g/mol
Melting Pt: 144…145 °C (dec.)
Density: 1,754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005637
CAS Number: 40432-84-6
EINECS: 254-917-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR