9-Chlorofluorene ≥98%
Supplier: Thermo Scientific
H50352.03
H50352.06
ALFAH50352.03EA
167
SGD
ALFAH50352.03
ALFAH50352.06
9-Chlorofluorene ≥98%
9-Chlorofluorene
Purity: 98%
CAS No.: 6630-65-5
Molecular Formula: C13H9Cl
Formula Weight: 200.67
Melting Point: 86-88°
MDL No.: MFCD00043468
Formula:
C₁₃H₉Cl MW: 200,67 g/mol Melting Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043468 CAS Number: 6630-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...