2-Chloro-4,6-difluoroaniline ≥98%
Supplier: Thermo Scientific
|
Danger
|
H50363.03
H50363.06
ALFAH50363.03EA
212
SGD
ALFAH50363.03
ALFAH50363.06
2-Chloro-4,6-difluoroaniline ≥98%
2-Chloro-4,6-difluoroaniline
Purity: 98%
CAS No.: 36556-56-6
Molecular Formula: C6H4ClF2N
Formula Weight: 163.55
Flash Point: 82°(180°F)
Melting Point: 28-32°
UN No.: 2811
MDL No.: MFCD00142892
Formula:
C₆H₄ClF₂N MW: 163,55 g/mol Melting Pt: 28…32 °C Flash Pt: 82 °C (180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142892 CAS Number: 36556-56-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...