4-Amino-6-chloropyrimidine 98%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Chloropyrimidin-4-ylamine, 6-Chloropyrimidin-4-amine, 6-Chloro-4-pyrimidinamine, 6-Chlor-4-pyrimidinamin

H50551.03 H50551.MD
ALFAH50551.03EA 126 SGD
ALFAH50551.03 ALFAH50551.MD
4-Amino-6-chloropyrimidine 98%
4-Amino-6-chloropyrimidine

Purity: 98% CAS No.: 5305-59-9 Molecular Formula: C4H4ClN3 Formula Weight: 129.55 MDL No.: MFCD00053576

Formula: C₄H₄ClN₃
MW: 129,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053576
CAS Number: 5305-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR