(2,4-Xylyloxy)acetic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (2,4-Dimethylphenoxy)acetic acid, 2-(2,4-Dimethylphenoxy)acetic acid

L01296.06 L01296.14
ALFAL01296.06EA 0 SGD
ALFAL01296.06 ALFAL01296.14
(2,4-Xylyloxy)acetic acid ≥98%
(2,4-Xylyloxy)acetic acid

Purity: 98+% CAS No.: 13334-49-1 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 139-143° Beilstein Reference: 2263832

Formula: (C₆H₃(CH₃)₂)OCH₂COOH
MW: 180,2 g/mol
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00014355
CAS Number: 13334-49-1
EINECS: 236-385-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR