(±)-1-Nonen-3-ol ≥98%
Supplier: Thermo Scientific
Warning
|
Synonyms:
(±)-Hexyl vinyl carbinol, 1-Nonen-3-ol
L06889.06
L06889.14
ALFAL06889.06EA
87.9
SGD
ALFAL06889.06
ALFAL06889.14
(±)-1-Nonen-3-ol ≥98%
(±)-1-Nonen-3-ol
Purity: 98%
CAS No.: 21964-44-3
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 84°(183°F)
Refractive Index: 1.4420
Boiling Point: 192-194°
Beilstein Reference: 1745976
MDL No.: MFCD00021953
Formula:
C₉H₁₈O MW: 142,24 g/mol Boiling Pt: 192…194 °C Density: 0,840 Flash Pt: 84 °C (183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021953 CAS Number: 21964-44-3 EINECS: 244-686-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...