7-Nitro-1H-indazole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

L07970.03 L07970.06 L07970.14
ALFAL07970.03EA 142 SGD
ALFAL07970.03 ALFAL07970.06 ALFAL07970.14
7-Nitro-1H-indazole ≥98%
7-Nitro-1H-indazole

Purity: 98% CAS No.: 2942-42-9 Molecular Formula: C7H5N3O2 Formula Weight: 163.14 Melting Point: 184-189° Beilstein Reference: 6809

Formula: C₇H₅N₃O₂
MW: 163,14 g/mol
Melting Pt: 184…189 °C
Density: 1,56 g/cm³ (293 °C)
Storage Temperature: Ambient
MDL Number: MFCD00022789
CAS Number: 2942-42-9
EINECS: 220-934-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR