1-(2-Bromophenyl)-2-thiourea ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: N-(2-Bromophenyl)thiourea, 2-Bromophenylthiourea, 1-(2-Bromophenyl)thiourea

L10098.03 L10098.06
ALFAL10098.03EA 67 SGD
ALFAL10098.03 ALFAL10098.06
1-(2-Bromophenyl)-2-thiourea ≥98%
1-(2-Bromophenyl)-2-thiourea

Purity: 98%
CAS No.: 5391-30-0
Molecular Formula: C7H7BrN2S
Formula Weight: 231.12
Odour: Odourless
Melting Point: 124-126°
Beilstein Reference: 2718427
UN No.: 2811
MDL No.: MFCD00041146

Formula: (C₆H₄Br)NHC(S)NH₂
MW: 231,12 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 130…132,5 °C
MDL Number: MFCD00041146
CAS Number: 5391-30-0
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR