4-Bromo-o-toluonitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Bromo-2-methylbenzonitrile

L10526.06 L10526.14 L10526.03
ALFAL10526.06EA 157 SGD
ALFAL10526.06 ALFAL10526.14 ALFAL10526.03
4-Bromo-o-toluonitrile ≥98%
4-Bromo-o-toluonitrile

Purity: 98% CAS No.: 67832-11-5 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 63-64° Beilstein Reference: 2574103 UN No.: 3439 MDL No.: MFCD00041461

Formula: C₈H₆BrN
MW: 196,05 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 63…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00041461
CAS Number: 67832-11-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR