(4-Fluorophenoxy)acetic hydrazide ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(4-Fluorophenoxy)acetohydrazide, (4-Fluorophenoxy)acetic acid hydrazide, (4-Fluorophenoxy)acetohydrazide

L10649.03 L10649.06
ALFAL10649.03EA 0 SGD
ALFAL10649.03 ALFAL10649.06
(4-Fluorophenoxy)acetic hydrazide ≥98%
(4-Fluorophenoxy)acetic hydrazide

Purity: 98+% CAS No.: 1737-62-8 Molecular Formula: C8H9FN2O2 Formula Weight: 184.17 Melting Point: 124-126° Beilstein Reference: 2526730

Formula: (C₆H₄F)OCH₂CONHNH₂
MW: 184.17 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00082651
CAS Number: 1737-62-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR