3-(Diethylamino)propyl isothiocyanate 97%

Supplier: Thermo Scientific
Danger

L10750.03 L10750.06
ALFAL10750.03EA 0 SGD
ALFAL10750.03 ALFAL10750.06
3-(Diethylamino)propyl isothiocyanate 97%
3-(Diethylamino)propyl isothiocyanate

Purity: 97% CAS No.: 2626-52-0 Molecular Formula: (CH3CH2)2NCH2CH2CH2NCS Formula Weight: 172.29 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5030 Odour: Irritating Boiling Point: 94-95°/3.5mm Beilstein Reference: 1757446 UN No.: 2922 MDL No.: MFCD00041133

Formula: C₈H₁₆N₂S
Boiling Pt: 94…95 °C (3,5 mmHg)
Density: 0,950
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00041133
CAS Number: 2626-52-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR