5-Ethyluracil ≥98%
Supplier: Thermo Scientific
Synonyms:
Ethyluracil, 5-Ethylpyrimidine-2,4(1H,3H)-dione
L10861.03
L10861.MD
ALFAL10861.03EA
0
SGD
ALFAL10861.03
ALFAL10861.MD
5-Ethyluracil ≥98%
5-Ethyluracil
Purity: 98+%
CAS No.: 4212-49-1
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Melting Point: 308-310°
Beilstein Reference: 119494
Formula:
C₆H₈N₂O₂ MW: 140,14 g/mol Melting Pt: 308…310 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079187 CAS Number: 4212-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...