3-Isothiocyanatobenzoic acid ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Carboxyphenyl isothiocyanate
L11322.03
L11322.06
ALFAL11322.03EA
170
SGD
ALFAL11322.03
ALFAL11322.06
3-Isothiocyanatobenzoic acid ≥97%
3-Isothiocyanatobenzoic acid
Purity: 97%
CAS No.: 2131-63-7
Molecular Formula: C8H5NO2S
Formula Weight: 179.20
Odour: Irritating
Melting Point: ca 165° dec.
Beilstein Reference: 2209003
UN No.: 2923
MDL No.: MFCD00041367
Formula:
C₈H₅NO₂S MW: 179,2 g/mol Boiling Pt: 336 °C (1013 hPa) Melting Pt: ∼ 165 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041367 CAS Number: 2131-63-7 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...