1,3,5-Trichloro-2-iodobenzene 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,4,6-Trichloroiodobenzene
L11975.06
L11975.03
ALFAL11975.06EA
257
SGD
ALFAL11975.06
ALFAL11975.03
1,3,5-Trichloro-2-iodobenzene 96%
1,3,5-Trichloro-2-iodobenzene
Purity: 96%
CAS No.: 6324-50-1
Molecular Formula: C6H2Cl3I
Formula Weight: 307.35
Melting Point: 52-57°
Beilstein Reference: 1868441
Formula:
C₆H₂Cl₃I MW: 307,34 g/mol Melting Pt: 52…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052857 CAS Number: 6324-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...