5-Amino-3,4-dimethylisoxazole 97%

Supplier: Thermo Scientific

Synonyms: 3,4-Dimethylisoxazol-5-amine, 3,4-Dimethylisoxazol-5-ylamine

L12191.09 L12191.04
ALFAL12191.09EA 0 SGD
ALFAL12191.09 ALFAL12191.04
5-Amino-3,4-dimethylisoxazole 97%
5-Amino-3,4-dimethylisoxazole

Purity: 97% CAS No.: 19947-75-2 Molecular Formula: C5H8N2O Formula Weight: 112.13 Melting Point: 118-120° Beilstein Reference: 108546

Formula: C₅H₈N₂O
MW: 112,13 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00046081
CAS Number: 19947-75-2
EINECS: 243-437-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR