5-(4-Bromophenyl)-1H-tetrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
L15181.03
L15181.MD
ALFAL15181.03EA
0
SGD
ALFAL15181.03
ALFAL15181.MD
5-(4-Bromophenyl)-1H-tetrazole ≥97%
5-(4-Bromophenyl)-1H-tetrazole
Purity: 97%
CAS No.: 50907-23-8
Molecular Formula: C7H5BrN4
Formula Weight: 225.05
Melting Point: ca 268° dec.
Beilstein Reference: 141235
Formula:
C₇H₅BrN₄ MW: 225,05 g/mol Melting Pt: ∼ 268 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00991874 CAS Number: 50907-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...