5-(3-Fluorophenyl)-1H-tetrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
L15558.03
L15558.MD
ALFAL15558.03EA
0
SGD
ALFAL15558.03
ALFAL15558.MD
5-(3-Fluorophenyl)-1H-tetrazole ≥97%
5-(3-Fluorophenyl)-1H-tetrazole
Purity: 97%
CAS No.: 50907-20-5
Molecular Formula: C7H5FN4
Formula Weight: 164.14
Melting Point: 142-144°
Beilstein Reference: 1104440
Formula:
C₇H₅FN₄ MW: 164,14 g/mol Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075702 CAS Number: 50907-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...