5-(4-Fluorophenyl)-1H-tetrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
L15559.03
L15559.MD
ALFAL15559.03EA
0
SGD
ALFAL15559.03
ALFAL15559.MD
5-(4-Fluorophenyl)-1H-tetrazole ≥97%
5-(4-Fluorophenyl)-1H-tetrazole
Purity: 97%
CAS No.: 50907-21-6
Molecular Formula: C7H5FN4
Formula Weight: 164.14
Melting Point: 208-212°
Beilstein Reference: 1100438
Formula:
C₇H₅FN₄ MW: 164,14 g/mol Melting Pt: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00478201 CAS Number: 50907-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...