5-(Trifluoromethyl)uracil ≥97%
Supplier: Thermo Fisher Scientific
Synonyms:
Trifluorothymine, 5-(Trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
L16196.03
L16196.MD
ALFAL16196.03EA
0
SGD
ALFAL16196.03
ALFAL16196.MD
5-(Trifluoromethyl)uracil ≥97%
5-(Trifluoromethyl)uracil
Purity: 97%
CAS No.: 54-20-6
Molecular Formula: C5H3F3N2O2
Formula Weight: 180.08
Melting Point: 251-254
Beilstein Reference: 612694
Formula:
C₅H₃F₃N₂O₂ MW: 180.09 g/mol Boiling Pt: 350 Melting Pt: 251…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006024 CAS Number: 54-20-6 EINECS: 200-197-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...