(3,3,3-Trifluoropropyl)trichlorosilane ≥97%
Supplier: Thermo Scientific
|
Danger
|
L16670.06
L16670.14
ALFAL16670.06EA
0
SGD
ALFAL16670.06
ALFAL16670.14
(3,3,3-Trifluoropropyl)trichlorosilane ≥97%
(3,3,3-Trifluoropropyl)trichlorosilane
Purity: 97%
CAS No.: 592-09-6
Molecular Formula: C3H4Cl3F3Si
Formula Weight: 231.51
Flash Point: 15°(59°F)
Refractive Index: 1.3860
Boiling Point: 113-114°
Beilstein Reference: 1748876
UN No.: 2985
MDL No.: MFCD00013602
Formula:
F₃CCH₂CH₂SiCl₃ Boiling Pt: 113…114 °C Density: 1,395 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013602 CAS Number: 592-09-6 EINECS: 209-744-2 UN: 2985 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...