1,2,5-Trichloro-3-iodobenzene ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,3,5-Trichloroiodobenzene
L17657.14
L17657.03
L17657.06
ALFAL17657.14EA
718
SGD
ALFAL17657.14
ALFAL17657.03
ALFAL17657.06
1,2,5-Trichloro-3-iodobenzene ≥97%
1,2,5-Trichloro-3-iodobenzene
Purity: 97%
CAS No.: 216393-66-7
Molecular Formula: C6H2Cl3I
Formula Weight: 307.35
Melting Point: 50-54°
Beilstein Reference: 3128014
Formula:
C₆H₂Cl₃I MW: 307,34 g/mol Melting Pt: 50…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318175 CAS Number: 216393-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...