3-Chloro-2-fluorobenzylamine 97%

Supplier: Thermo Scientific
Danger

Synonyms: (3-Chloro-2-fluorophenyl)methanamine

L19227.03 L19227.MD
ALFAL19227.03EA 0 SGD
ALFAL19227.03 ALFAL19227.MD
3-Chloro-2-fluorobenzylamine 97%
3-Chloro-2-fluorobenzylamine

Purity: 97% CAS No.: 72235-55-3 Molecular Formula: C7H7ClFN Formula Weight: 159.59 Refractive Index: 1.5405

Formula: C₇H₇ClFN
MW: 159,59 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD01631391
CAS Number: 72235-55-3
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR