2-Amino-5-chloropyrimidine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Chloropyrimidin-2-ylamine, 5-Chloro-2-pyrimidinamine, 5-Chlor-2-pyrimidinamin, 5-chlorpyrimidin-2-amin

L19248.03 L19248.06 L19248.MD
ALFAL19248.03EA 0 SGD
ALFAL19248.03 ALFAL19248.06 ALFAL19248.MD
2-Amino-5-chloropyrimidine 97%
2-Amino-5-chloropyrimidine

Purity: 97% CAS No.: 5428-89-7 Molecular Formula: C4H4ClN3 Formula Weight: 129.55 Melting Point: 235° Beilstein Reference: 110914

Formula: C₄H₄ClN₃
MW: 129,55 g/mol
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00014604
CAS Number: 5428-89-7
EINECS: 226-581-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR