2-Amino-4-(trifluoromethyl)pyridine 99%
Supplier: Thermo Scientific
|
Danger
|
L19548.MC
L19548.ME
ALFAL19548.MCEA
0
SGD
ALFAL19548.MC
ALFAL19548.ME
2-Amino-4-(trifluoromethyl)pyridine 99%
2-Amino-4-(trifluoromethyl)pyridine
Purity: 99%
CAS No.: 106447-97-6
Molecular Formula: C6H5F3N2
Formula Weight: 162.11
Melting Point: 71-74°
Beilstein Reference: 7869677
UN No.: 2811
MDL No.: MFCD00077455
Formula:
C₆H₅F₃N₂ MW: 162,11 g/mol Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077455 CAS Number: 106447-97-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...