4-Amino-3-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific
Danger

L19549.MC L19549.ME
ALFAL19549.MCEA 0 SGD
ALFAL19549.MC ALFAL19549.ME
4-Amino-3-(trifluoromethyl)pyridine 97%
4-Amino-3-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 387824-61-5 Molecular Formula: C6H5F3N2 Formula Weight: 162.11 Melting Point: 98-102° UN No.: 2811 MDL No.: MFCD02183554

Formula: C₆H₅F₃N₂
MW: 162,11 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD02183554
CAS Number: 387824-61-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR