2-Aminonicotinonitrile 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine
L19618.03
L19618.06
ALFAL19618.03EA
60.6
SGD
ALFAL19618.03
ALFAL19618.06
2-Aminonicotinonitrile 98%
2-Aminonicotinonitrile
Purity: 98%
CAS No.: 24517-64-4
Molecular Formula: C6H5N3
Formula Weight: 119.13
Melting Point: 133-135°
Beilstein Reference: 115612
UN No.: 3439
MDL No.: MFCD00128867
Formula:
C₆H₅N₃ MW: 119,13 g/mol Boiling Pt: 268 °C (1013 hPa) Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128867 CAS Number: 24517-64-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...