3,5-Difluoropyridine-2-carbonitrile 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3,5-Difluoropicolinonitrile, 2-Cyano-3,5-difluoropyridine

L19724.03 L19724.06
ALFAL19724.03EA 0 SGD
ALFAL19724.03 ALFAL19724.06
3,5-Difluoropyridine-2-carbonitrile 98
3,5-Difluoropyridine-2-carbonitrile

Purity: 98% CAS No.: 298709-29-2 Molecular Formula: C6H2F2N2 Formula Weight: 140.09 Melting Point: 30-34° MDL No.: MFCD03788759

Formula: C₆H₂F₂N₂
MW: 140.09 g/mol
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD03788759
CAS Number: 298709-29-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR