2,6-Dichloro-3-(trifluoromethyl)pyridine ≥98%

Supplier: Thermo Scientific
Danger

L20003.03 L20003.MD
ALFAL20003.03EA 453 SGD
ALFAL20003.03 ALFAL20003.MD
2,6-Dichloro-3-(trifluoromethyl)pyridine ≥98%
2,6-Dichloro-3-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 55304-75-1 Molecular Formula: C6H2Cl2F3N Formula Weight: 215.99 Refractive Index: 1.4845 Boiling Point: 194-196° UN No.: 2810 MDL No.: MFCD00042245

Formula: C₆H₂Cl₂F₃N
MW: 215,99 g/mol
Boiling Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00042245
CAS Number: 55304-75-1
EINECS: 259-585-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR