2-Acetyl-3-bromothiophene ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Acetyl-3-boromothiophene, 1-(3-Bromo-2-thienyl)-1-ethanone
L20113.03
L20113.06
ALFAL20113.03EA
0
SGD
ALFAL20113.03
ALFAL20113.06
2-Acetyl-3-bromothiophene ≥97%
2-Acetyl-3-bromothiophene
Purity: 97%
CAS No.: 42877-08-7
Molecular Formula: C6H5BrOS
Formula Weight: 205.07
Refractive Index: 1.6140
Melting Point: 41-45°
Boiling Point: 102-105°/4mm
Beilstein Reference: 116973
MDL No.: MFCD00794287
Formula:
C₆H₅BrOS MW: 205,08 g/mol Boiling Pt: 102…105 °C (4 mmHg) Melting Pt: 41…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00794287 CAS Number: 42877-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...