2-Acetyl-3-chlorothiophene ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(3-Chloro-2-thienyl)-1-ethanone

L20114.03 L20114.06 L20114.14
ALFAL20114.03EA 87 SGD
ALFAL20114.03 ALFAL20114.06 ALFAL20114.14
2-Acetyl-3-chlorothiophene ≥97%
2-Acetyl-3-chlorothiophene

Purity: 97% CAS No.: 89581-82-8 Molecular Formula: C6H5ClOS Formula Weight: 160.62 Refractive Index: 1.5840 Boiling Point: 117-118°/15mm UN No.: 2810 MDL No.: MFCD00794248

Formula: C₆H₅ClOS
MW: 160,62 g/mol
Boiling Pt: 117…118 °C (15 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00794248
CAS Number: 89581-82-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR