Methanol ≥99.9%, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

NP230-10 NP230-20 230-4
BJAANP230-10EA 1840 SGD
BJAANP230-10 BJAANP230-20 BJAA230-4
Methanol ≥99.9%, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Methanol

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃OH
Boiling Pt: 64,7 °C
Melting Pt: –98 °C
Density: 0,792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=7d8032d09939fbf669cffa2c324454ca9267723e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Fluorescence (as quinine base) 0.30 ppb max.
GC Analysis 99.9 % min.
HPLC Gradient @ 220 nm 0.005 AU max.
HPLC Gradient @ 254 nm 0.001 AU max.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
UV Cutoff 205 nm max.
Water by Karl Fischer Titration 0.05 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR