Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

NP407-20 407-4
BJAANP407-20EA 8570 SGD
BJAANP407-20 BJAA407-4
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile with 0.1% (v/v) trifluoroacetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C (1,013 hPa)
Melting Pt: –46 °C
Density: 0,777 g/cm³ (25 °C)
Flash Pt: 6 °C (main component)
Storage Temperature: Ambient
EINECS: 200-929-3
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Trifluoroacetic Acid (v/v) 0.09 - 0.11%
UV Cutoff <190 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 215 nm < 0.020 AU
HPLC Gradient @ 254 nm < 0.010 AU
UV Absorbance @ 210 nm < 0.360 AU
UV Absorbance @ 230 nm < 0.250 AU
UV Absorbance @ 250 nm < 0.040 AU
UV Absorbance @ 300 nm < 0.005 AU
UV Absorbance @ 400 nm < 0.005 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR