Acetonitrile with 0,05% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger


Stabilization: Stabilised with BHT (Ionol) 250 ppm

NP423-20 423-4
BJAANP423-20EA 5730 SGD
BJAANP423-20 BJAA423-4
Acetonitrile with 0,05% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Acetonitrile with 0,05% (v/v) trifluoroacetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C (1,013 hPa)
Melting Pt: –46 °C
Density: 0,777 g/cm³ (25 °C)
Flash Pt: 6 °C (main component)
Storage Temperature: Ambient
EINECS: 200-929-3
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Trifluoroacetic Acid (v/v) 0.045 - 0.055%
UV Cutoff <189 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 215 nm < 0.020 AU
HPLC Gradient @ 254 nm < 0.010 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR