Dimethyl sulphoxide, anhydrous ≥99.8% for applications requiring low water content, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Warning

AS081-0100 AS081-1
BJAAAS081-0100EA 843 SGD
BJAAAS081-0100 BJAAAS081-1
Dimethyl sulphoxide, anhydrous ≥99.8% for applications requiring low water content, Burdick & Jackson™
Dimethyl sulphoxide

Septum-sealed bottle


  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

Formula: (CH₃)₂SO
Boiling Pt: 189 °C
Melting Pt: 18,5 °C
Density: 1,0954 g/cm3
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=ef2c8190e048406b967644856c8e2e7172d77412&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Color (APHA) 10 max.
GC Analysis 99.8 % min.
Residue 5 mg/L max.
Water by Karl Fischer Titration 50 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR