Methanol, anhydrous ≥99.9% for applications requiring low water content, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

Synonyms: Hydroxy methane, Carbinol, Wood alcohol, Methyl alcohol

AS230-0100 AS230-1
BJAAAS230-0100EA 607 SGD
BJAAAS230-0100 BJAAAS230-1
Methanol, anhydrous ≥99.9% for applications requiring low water content, Burdick & Jackson™
Methanol

Septum-sealed bottle


  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

Formula: CH₃OH
MW: 32,04 g/mol
Boiling Pt: 64,7 °C
Melting Pt: –98 °C
Density: 0,792 g/cm3
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=ee3ca6f8de15ff99d2c30d397ca2bb30164bfae5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Color (APHA) 10 max.
GC Analysis 99.9 % min.
Residue 3 mg/L max.
Water by Karl Fischer Titration 50 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR