Tetrahydrofuran, anhydrous ≥99.9% contains no preservatives for applications requiring low water content, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

AS340-0100 AS340-1
BJAAAS340-0100EA 898 SGD
BJAAAS340-0100 BJAAAS340-1
Tetrahydrofuran, anhydrous ≥99.9% contains no preservatives for applications requiring low water content, Burdick & Jackson™
Tetrahydrofuran

Septum-sealed bottle


  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

Formula: C₄H₈O
MW: 72,11 g/mol
Boiling Pt: 66 °C
Melting Pt: –108,5 °C
Density: 0,8892 g/cm3
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=7e2e27ffa486f0bd05a19c93df23d5cd06e9ad9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Color (APHA) 10 max.
GC Analysis 99.9 % min.
Peroxides (as H₂O₂) at time of packaging 10 ppm max.
Residue 3 mg/L max.
Water by Karl Fischer Titration 50 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR