Triethylamine ≥99.5%, BioSyn™ for biosynthesis, for HPLC, Fluka™

Supplier: BURDICK & JACKSON
Danger

Synonyms: (Diethylamino)ethane, TEA

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm

BB359-1 BB359P100
BJAABB359-1EA 2750 SGD
BJAABB359-1 BJAABB359P100
Triethylamine ≥99.5%, BioSyn™ for biosynthesis, for HPLC, Fluka™
Triethylamine

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: (C₂H₅)₃N
MW: 101,19 g/mol
Boiling Pt: 89,5 °C
Melting Pt: –115 °C
Density: 0,729 g/cm3
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=47941a0f453629002c367c247ada0357203ff866&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
GC Analysis 99.5 % min.
Water by Karl Fischer Titration 0.2 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR