Chloroform ≥99.8% stabilised ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: BURDICK & JACKSON
Danger


Stabilization: Stabilised with amylene

LP049-4 LP049TC19
BJAALP049-4EA 1660 SGD
BJAALP049-4 BJAALP049TC19
Chloroform ≥99.8% stabilised ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Chloroform

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: CHCl₃
Boiling Pt: 62 °C
Melting Pt: –64 °C
Density: 1,49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.8 % min.
Color (APHA) 10 max.
Free Chlorine Passes test
Lead 0.05 ppm max.
Residue after Evaporation 0.001 % max.
Water by Karl Fischer Titration 0.02 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR