Heptane (mixture of isomers) ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: BURDICK & JACKSON
Danger

LP206-4 LP206D19
BJAALP206-4EA 1480 SGD
BJAALP206-4 BJAALP206D19
Heptane (mixture of isomers) ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Heptane (mixture of isomers)

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: H₃C(CH₂)₅CH₃
MW: 100,2 g/mol
Boiling Pt: 98,4 °C
Melting Pt: -90,6 °C
Density: 0,684 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=b57298f7f7280e366488444db9c9b34698895b06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis (n-Heptane) 99 % min.
Assay by GC Analysis (total C7 isomers) 99.9 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.001 % max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR