1-Propanol ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: BURDICK JACKSON
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

LP322-4 LP322D19
BJAALP322-4EA 3720 SGD
BJAALP322-4 BJAALP322D19
1-Propanol ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
1-Propanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: H₃CCH₂CH₂OH
MW: 60,1 g/mol
Boiling Pt: 13,7 %(V)
Melting Pt: -126,2 °C
Density: 0,8053 g/cm³ (20 °C)
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,III
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.5 % min.
Water by Karl Fischer Titration 0.2 % max.
Carbonyl Compounds (as C2H5CHO) 0.03 % max.
Color (APHA) 10 max.
Ethyl Alcohol 0.01 % max.
Isopropyl Alcohol 0.05 % max.
Methanol 0.01 % max.
Residue after Evaporation 0.001 % max.
Solubility in Water Passes test
Titrable Acid 0.4 µeq/g max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR