2,2,4-Trimethylpentane ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: BURDICK & JACKSON
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

LP362-4 LP362D19
BJAALP362-4EA 2940 SGD
BJAALP362-4 BJAALP362D19
2,2,4-Trimethylpentane ≥99.0% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
2,2,4-Trimethylpentane

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114,23 g/mol
Boiling Pt: 99,24 °C
Melting Pt: –107,4 °C
Density: 0,69 g/cm³ (20 °C)
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=f42ff6c045f5e1bfc46620eb239a7a8f6b987217&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99 % min.
Color (APHA) 10 max.
Residue after Evaporation 5 mg/L max.
Sulfur Compounds (as S) 0.005 % max.
Water by Karl Fischer Titration 0.01 % max.
Water-Soluble Titrable Acid 0.3 µeq/g max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR