Methyl ethyl ketone ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Supplier: BURDICK JACKSON
|
Danger
|
Synonyms:
EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
NP247-20
247-4
BJAANP247-20EA
2490
SGD
BJAANP247-20
BJAA247-4
Methyl ethyl ketone ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Methyl ethyl ketone
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
C₂H₅COCH₃ MW: 72.11 g/mol Boiling Pt: 79.64 °C Melting Pt: –86.69 °C Density: 0.805 g/cm³ (20 °C) Flash Pt: –9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3 II REACH: 01-2119457290-43 Merck Index: 13,06097 |
|
Specification Test Results
GC Analysis | 99.5 % min. |
Refractive Index (20ºC) | 1.3776 - 1.3790 |
Residue | 3 mg/L max. |
UV Absorbance @ 340 nm | 0.1 AU max. |
UV Absorbance @ 350 nm | 0.02 AU max. |
UV Absorbance @ 375 nm | 0.01 AU max. |
UV Absorbance @ 400 nm | 0.005 AU max. |
UV Cutoff | 329 nm max. |
Water by Karl Fischer Titration | 0.05 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...