2,2,4-Trimethylpentane ≥99.0%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

NP362-20 362-4
BJAANP362-20EA 2350 SGD
BJAANP362-20 BJAA362-4
2,2,4-Trimethylpentane ≥99.0%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
2,2,4-Trimethylpentane

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99,24 °C
Melting Pt: -107,4 °C
Density: 0,69 g/cm³ (20 °C)
Flash Pt: -8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=559f761f651e5fcdb0cf5dd303cd79e53daf2366&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 2 µg/L max.
GC Analysis 99 % min.
Refractive Index (20ºC) 1.3907 - 1.3923
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.1 AU max.
UV Absorbance @ 250 nm 0.02 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Cutoff 215 nm max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR