Ethyl acetate ≥99.5%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

PP100-19 PP100-204
BJAAPP100-19EA 1940 SGD
BJAAPP100-19 BJAAPP100-204
Ethyl acetate ≥99.5%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Ethyl acetate

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: CH₃COOC₂H₅
Boiling Pt: 11,5 %(V)
Melting Pt: –84 °C
Density: 0,9006 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.9 % min.
Color (APHA) 10 max.
Refractive Index (20ºC) 1.3713 - 1.3733
Residue after Evaporation 5 mg/L max.
Substances Darkened by Sulfuric Acid Passes test
Titrable Acid 0.9 µeq/g max.
Water by Karl Fischer Titration 0.1 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR