1-Propanol ≥99.5%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK & JACKSON
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

PP322-19 PP322-204
BJAAPP322-19EA 1710 SGD
BJAAPP322-19 BJAAPP322-204
1-Propanol ≥99.5%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
1-Propanol

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: H₃CCH₂CH₂OH
MW: 60,1 g/mol
Boiling Pt: 97,2 °C
Melting Pt: –126,2 °C
Density: 0,8053 g/cm3
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,III
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=402731a24fb391e661ea44080e5d9a62e0a25788&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.5 % min.
Water by Karl Fischer Titration 0.1 % max.
Carbonyl Compounds (as C2H5CHO) 0.03 % max.
Color (APHA) 10 max.
Ethyl Alcohol 0.01 % max.
Isopropyl Alcohol 0.05 % max.
Methanol 0.01 % max.
Refractive Index (20ºC) 1.3839 - 1.3859
Residue after Evaporation 5 mg/L max.
Titrable Acid 0.4 µeq/g max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR